Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC23
Entrez Gene ID
64927
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
HCC-8
Chromosome Location
Chr.15: 99136323 - 99251226 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288615.1 NP_001275544.1 8 - 9 1459 133
NM_001288616.1 NP_001275545.1 6 - 7 850 133
NR_109947.1 7 - 8 1120 133
XM_011521927.2 XP_011520229.1 4 - 5 600 133
XM_011521929.1 XP_011520231.1 8 - 9 1167 133
XM_011521930.1 XP_011520232.1 7 - 8 1045 133
XM_011521931.1 XP_011520233.1 6 - 7 1188 133
XM_011521932.1 XP_011520234.1 7 - 8 1056 133
XM_011521933.1 XP_011520235.1 6 - 7 989 133
XM_011521934.1 XP_011520236.1 8 - 9 1459 133
XM_011521935.1 XP_011520237.1 6 - 7 1171 133
XM_011521936.1 XP_011520238.1 6 - 7 850 133
XM_011521937.1 XP_011520239.1 5 - 6 1040 133
XM_011521939.1 XP_011520241.1 6 - 7 916 133
XM_011521940.2 XP_011520242.1 4 - 5 600 133
XM_011521941.2 XP_011520243.1 4 - 5 600 133
XM_011521943.2 XP_011520245.1 4 - 5 600 133
XM_011521944.2 XP_011520246.1 4 - 5 600 133
XM_011521945.2 XP_011520247.1 4 - 5 600 133
XM_017022509.1 XP_016877998.1 8 - 9 1618 133
XM_017022510.1 XP_016877999.1 7 - 8 1281 133
XM_017022511.1 XP_016878000.1 6 - 7 1055 133
XM_017022512.1 XP_016878001.1 6 - 7 1188 133
XM_017022513.1 XP_016878002.1 7 - 8 1056 133
XM_017022514.1 XP_016878003.1 6 - 7 989 133
XM_017022515.1 XP_016878004.1 6 - 7 1171 133
XM_017022516.1 XP_016878005.1 6 - 7 1055 133
XM_017022517.1 XP_016878006.1 7 - 8 972 133
XM_017022518.1 XP_016878007.1 6 - 7 916 133
GenBank mRNA AF411456.1 - 8 - 9 1430 133
AK023001.1 - 3 - 4 602 133
AK023230.1 - 8 - 9 1158 133
AK291690.1 - 6 - 7 847 133
BC015728.1 - 7 - 8 1019 133
DA681018.1 - 2 - 3 191 133

View Full Product Details