The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101667.1 | NP_001095137.1 | 6 - 7 | 777 | 71 |
NM_003501.2 | NP_003492.2 | 6 - 7 | 777 | 71 | |
XM_005248011.4 | XP_005248068.1 | 6 - 7 | 830 | 71 | |
XM_005248013.3 | XP_005248070.1 | 6 - 7 | 830 | 71 | |
XM_011513565.2 | XP_011511867.1 | 6 - 7 | 789 | 71 | |
XM_011513566.1 | XP_011511868.1 | 6 - 7 | 830 | 71 | |
GenBank mRNA | AK225080.1 | - | 6 - 7 | 775 | 71 |
AK313242.1 | - | 6 - 7 | 830 | 71 | |
BC017053.1 | - | 6 - 7 | 763 | 71 | |
Y11411.1 | - | 6 - 7 | 755 | 71 |