The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006624.1 | NP_001006625.1 | 1 - 2 | 171 | 84 |
NM_001006625.1 | NP_001006626.1 | 1 - 2 | 171 | 84 |