The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024870.3 | NP_079146.2 | 17 - 18 | 2164 | 168 |
NM_025170.5 | NP_079446.3 | 17 - 18 | 2164 | 168 | |
XM_011517612.2 | XP_011515914.1 | 17 - 18 | 2358 | 168 | |
GenBank mRNA | AJ437636.2 | - | 17 - 18 | 2161 | 168 |
AK023049.1 | - | 12 - 13 | 1239 | 168 | |
AK024079.1 | - | 2 - 3 | 240 | 168 | |
AK294299.1 | - | 15 - 16 | 1595 | 168 | |
AY508996.1 | - | 17 - 18 | 1911 | 168 | |
AY508997.1 | - | 17 - 18 | 1911 | 168 |