The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024870.3 | NP_079146.2 | 22 - 23 | 2832 | 141 |
NM_025170.5 | NP_079446.3 | 22 - 23 | 2832 | 141 | |
XM_011517612.2 | XP_011515914.1 | 22 - 23 | 3026 | 141 | |
GenBank mRNA | AJ437636.2 | - | 22 - 23 | 2829 | 141 |
AK023049.1 | - | 17 - 18 | 1907 | 141 | |
AK024079.1 | - | 7 - 8 | 908 | 141 | |
AK294299.1 | - | 20 - 21 | 2263 | 141 | |
AY508996.1 | - | 22 - 23 | 2579 | 141 | |
AY508997.1 | - | 22 - 23 | 2579 | 141 |