The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024870.3 | NP_079146.2 | 9 - 10 | 1372 | 90 |
NM_025170.5 | NP_079446.3 | 9 - 10 | 1372 | 90 | |
XM_011517612.2 | XP_011515914.1 | 9 - 10 | 1566 | 90 | |
GenBank mRNA | AJ437636.2 | - | 9 - 10 | 1369 | 90 |
AK023049.1 | - | 4 - 5 | 447 | 90 | |
AK294299.1 | - | 7 - 8 | 803 | 90 | |
AY508996.1 | - | 9 - 10 | 1119 | 90 | |
AY508997.1 | - | 9 - 10 | 1119 | 90 |