The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004447.5 | NP_004438.3 | 15 - 16 | 1996 | 98 |
XM_011520605.2 | XP_011518907.1 | 16 - 17 | 2056 | 98 | |
GenBank mRNA | AK291777.1 | - | 15 - 16 | 1801 | 98 |
AK292931.1 | - | 15 - 16 | 1653 | 98 | |
AK301834.1 | - | 8 - 9 | 986 | 98 | |
AK304860.1 | - | 15 - 16 | 1996 | 98 | |
AK316134.1 | - | 8 - 9 | 905 | 98 | |
AK316239.1 | - | 8 - 9 | 1178 | 98 | |
BC030010.2 | - | 15 - 16 | 1808 | 98 | |
U12535.1 | - | 15 - 16 | 1768 | 98 |