The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014824.2 | NP_055639.2 | 6 - 7 | 737 | 105 |
XM_011545409.1 | XP_011543711.1 | 6 - 7 | 619 | 105 | |
XM_011545410.2 | XP_011543712.1 | 6 - 7 | 552 | 105 | |
XM_017018632.1 | XP_016874121.1 | 6 - 7 | 796 | 105 | |
GenBank mRNA | AB018312.1 | - | 5 - 6 | 587 | 105 |
AK296376.1 | - | 5 - 6 | 407 | 105 | |
AK297928.1 | - | 6 - 7 | 621 | 105 | |
AL133567.1 | - | 1 - 2 | 105 | 105 | |
BC010394.1 | - | 6 - 7 | 484 | 105 |