The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205319.1 | NP_001192248.1 | 3 - 4 | 356 | 88 |
NM_022463.4 | NP_071908.2 | 3 - 4 | 699 | 88 | |
XM_005256756.4 | XP_005256813.1 | 3 - 4 | 705 | 88 | |
XM_005256758.3 | XP_005256815.1 | 3 - 4 | 1028 | 88 | |
XM_017024949.1 | XP_016880438.1 | 3 - 4 | 707 | 88 | |
GenBank mRNA | AK022676.1 | - | 1 - 2 | 128 | 88 |
AK027451.1 | - | 2 - 3 | 252 | 88 | |
AK302073.1 | - | 3 - 4 | 358 | 88 | |
AW752727.1 | - | 3 - 4 | 288 | 88 | |
AY189690.1 | - | 2 - 3 | 239 | 88 | |
BC009327.2 | - | 3 - 4 | 293 | 88 | |
BC073845.1 | - | 1 - 2 | 109 | 88 | |
BC104634.1 | - | 3 - 4 | 392 | 88 | |
CX786695.1 | - | 3 - 4 | 699 | 88 |