Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DEF8
- Entrez Gene ID
-
54849
- Gene Name
- differentially expressed in FDCP 8 homolog (mouse)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 89948697 - 89968060 on Build GRCh38
View Full Product Details