The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286771.2 | NP_001273700.1 | 19 - 20 | 3375 | 89 |
NM_015574.1 | NP_056389.1 | 19 - 20 | 3754 | 89 | |
NM_032217.4 | NP_115593.3 | 19 - 20 | 3757 | 89 | |
NM_198889.2 | NP_942592.1 | 18 - 19 | 3004 | 89 | |
XM_005265671.3 | XP_005265728.1 | 18 - 19 | 3345 | 89 | |
XM_005265672.3 | XP_005265729.1 | 19 - 20 | 4101 | 89 | |
XM_005265673.3 | XP_005265730.1 | 19 - 20 | 4101 | 89 | |
XM_017008011.1 | XP_016863500.1 | 19 - 20 | 3329 | 89 | |
XM_017008012.1 | XP_016863501.1 | 18 - 19 | 2579 | 89 | |
XM_017008013.1 | XP_016863502.1 | 18 - 19 | 2576 | 89 | |
XM_017008014.1 | XP_016863503.1 | 19 - 20 | 4098 | 89 | |
XM_017008015.1 | XP_016863504.1 | 19 - 20 | 3332 | 89 | |
XM_017008016.1 | XP_016863505.1 | 18 - 19 | 3348 | 89 | |
XM_017008017.1 | XP_016863506.1 | 18 - 19 | 2579 | 89 | |
GenBank mRNA | AB014597.2 | - | 19 - 20 | 3278 | 89 |
AK025859.1 | - | 4 - 5 | 527 | 89 | |
AK055209.1 | - | 11 - 12 | 1232 | 89 | |
AK299046.1 | - | 13 - 14 | 2257 | 89 | |
AK300302.1 | - | 18 - 19 | 3133 | 89 | |
AK307581.1 | - | 18 - 19 | 3004 | 89 | |
AL831903.1 | - | 7 - 8 | 1557 | 89 | |
AL833287.1 | - | 15 - 16 | 2181 | 89 | |
BC004173.2 | - | 19 - 20 | 3738 | 89 | |
BC007747.2 | - | 17 - 18 | 2858 | 89 | |
BC029935.2 | - | 2 - 3 | 282 | 89 | |
BX648035.1 | - | 19 - 20 | 3330 | 89 |