The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014153.3 | NP_054872.2 | 18 - 19 | 2386 | 146 |
XM_006720877.2 | XP_006720940.1 | 18 - 19 | 2422 | 146 | |
XM_011522463.1 | XP_011520765.1 | 19 - 20 | 2571 | 146 | |
XM_011522464.2 | XP_011520766.1 | 12 - 13 | 2179 | 146 | |
XM_017023174.1 | XP_016878663.1 | 18 - 19 | 2403 | 146 | |
XM_017023175.1 | XP_016878664.1 | 14 - 15 | 2016 | 146 | |
GenBank mRNA | AK000325.1 | - | 16 - 17 | 2125 | 146 |
AK026956.1 | - | 10 - 11 | 1611 | 146 | |
AK091803.1 | - | 16 - 17 | 2088 | 146 | |
AY163807.1 | - | 17 - 18 | 2415 | 146 | |
BC046363.1 | - | 18 - 19 | 2279 | 146 |