The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014153.3 | NP_054872.2 | 19 - 20 | 2492 | 107 |
XM_006720877.2 | XP_006720940.1 | 19 - 20 | 2528 | 107 | |
XM_011522463.1 | XP_011520765.1 | 20 - 21 | 2677 | 107 | |
XM_011522464.2 | XP_011520766.1 | 13 - 14 | 2285 | 107 | |
XM_017023174.1 | XP_016878663.1 | 19 - 20 | 2509 | 107 | |
XM_017023175.1 | XP_016878664.1 | 15 - 16 | 2122 | 107 | |
GenBank mRNA | AK000325.1 | - | 17 - 18 | 2231 | 107 |
AK026956.1 | - | 11 - 12 | 1717 | 107 | |
AK091803.1 | - | 17 - 18 | 2194 | 107 | |
AY163807.1 | - | 18 - 19 | 2521 | 107 | |
BC046363.1 | - | 19 - 20 | 2385 | 107 |