Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H7A
Entrez Gene ID
29066
Gene Name
zinc finger CCCH-type containing 7A
Gene Aliases
HSPC055, ZC3H7, ZC3HDC7
Chromosome Location
Chr.16: 11750586 - 11797258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014153.3 NP_054872.2 6 - 7 720 130
XM_006720877.2 XP_006720940.1 6 - 7 756 130
XM_011522463.1 XP_011520765.1 7 - 8 905 130
XM_017023174.1 XP_016878663.1 6 - 7 737 130
XM_017023175.1 XP_016878664.1 3 - 4 415 130
GenBank mRNA AK000325.1 - 4 - 5 460 130
AK091803.1 - 6 - 7 692 130
AY163807.1 - 5 - 6 749 130
BC036857.1 - 6 - 7 664 130
BC046363.1 - 6 - 7 613 130

View Full Product Details