Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAMP
Entrez Gene ID
14
Gene Name
angio associated migratory cell protein
Gene Aliases
-
Chromosome Location
Chr.2: 218264129 - 218270209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001087.4 NP_001078.2 6 - 7 889 132
NM_001302545.1 NP_001289474.1 6 - 7 892 132
GenBank mRNA AK131047.1 - 6 - 7 826 132
AK225078.1 - 6 - 7 797 132
BC008809.2 - 3 - 4 372 132
BC020244.1 - 6 - 7 753 132
BC039866.1 - 6 - 7 817 132
CR456755.1 - 6 - 7 820 132
M95627.1 - 6 - 7 820 132

View Full Product Details