Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
Chromosome Location
Chr.7: 91940867 - 92110673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005751.4 NP_005742.4 25 - 26 6438 157
NM_147185.2 NP_671714.1 25 - 26 6438 157
XM_017011642.1 XP_016867131.1 27 - 28 6581 157
XM_017011643.1 XP_016867132.1 26 - 27 6542 157
XM_017011644.1 XP_016867133.1 27 - 28 6581 157
XM_017011645.1 XP_016867134.1 26 - 27 6527 157
XM_017011646.1 XP_016867135.1 26 - 27 6542 157
XM_017011647.1 XP_016867136.1 26 - 27 6488 157
XM_017011648.1 XP_016867137.1 26 - 27 6485 157
XM_017011649.1 XP_016867138.1 26 - 27 6542 157
XM_017011650.1 XP_016867139.1 25 - 26 6446 157
XM_017011651.1 XP_016867140.1 26 - 27 6440 157
XM_017011652.1 XP_016867141.1 27 - 28 6581 157
XM_017011653.1 XP_016867142.1 24 - 25 6353 157
XM_017011654.1 XP_016867143.1 24 - 25 6305 157
XM_017011655.1 XP_016867144.1 27 - 28 6240 157
XM_017011656.1 XP_016867145.1 26 - 27 6170 157
XM_017011657.1 XP_016867146.1 12 - 13 2090 157
XM_017011658.1 XP_016867147.1 5 - 6 1452 157
XM_017011659.1 XP_016867148.1 4 - 5 1413 157
XM_017011660.1 XP_016867149.1 4 - 5 1413 157
GenBank mRNA AB019691.1 - 25 - 26 6299 157
AF083037.1 - 18 - 19 5277 157
AF247727.4 - 24 - 25 6072 157
AJ131693.1 - 25 - 26 6438 157
AY528715.1 - 7 - 8 1285 157

View Full Product Details