Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
Chromosome Location
Chr.7: 91940867 - 92110673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005751.4 NP_005742.4 43 - 44 10944 72
NM_147185.2 NP_671714.1 43 - 44 10920 72
XM_017011642.1 XP_016867131.1 45 - 46 11087 72
XM_017011643.1 XP_016867132.1 44 - 45 11048 72
XM_017011644.1 XP_016867133.1 45 - 46 11087 72
XM_017011645.1 XP_016867134.1 44 - 45 11033 72
XM_017011646.1 XP_016867135.1 44 - 45 11048 72
XM_017011647.1 XP_016867136.1 44 - 45 10994 72
XM_017011648.1 XP_016867137.1 44 - 45 10991 72
XM_017011649.1 XP_016867138.1 44 - 45 11024 72
XM_017011650.1 XP_016867139.1 43 - 44 10952 72
XM_017011651.1 XP_016867140.1 44 - 45 10946 72
XM_017011652.1 XP_016867141.1 44 - 45 10898 72
XM_017011653.1 XP_016867142.1 42 - 43 10859 72
XM_017011654.1 XP_016867143.1 42 - 43 10811 72
XM_017011655.1 XP_016867144.1 45 - 46 10746 72
XM_017011656.1 XP_016867145.1 44 - 45 10676 72
XM_017011657.1 XP_016867146.1 30 - 31 6596 72
XM_017011658.1 XP_016867147.1 23 - 24 5958 72
XM_017011659.1 XP_016867148.1 22 - 23 5919 72
XM_017011660.1 XP_016867149.1 22 - 23 5919 72
GenBank mRNA AB018346.1 - 16 - 17 4123 72
AB019691.1 - 43 - 44 10781 72
AF083037.1 - 36 - 37 9783 72
AF091711.1 - 17 - 18 4264 72
AJ131693.1 - 43 - 44 10944 72
AK025160.1 - 3 - 4 563 72
AK026444.1 - 5 - 6 1643 72
AK074869.1 - 13 - 14 2823 72
BC040932.1 - 2 - 3 318 72

View Full Product Details