The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005751.4 | NP_005742.4 | 45 - 46 | 11314 | 80 |
NM_147185.2 | NP_671714.1 | 45 - 46 | 11290 | 80 | |
XM_017011642.1 | XP_016867131.1 | 47 - 48 | 11457 | 80 | |
XM_017011643.1 | XP_016867132.1 | 46 - 47 | 11418 | 80 | |
XM_017011644.1 | XP_016867133.1 | 47 - 48 | 11457 | 80 | |
XM_017011645.1 | XP_016867134.1 | 46 - 47 | 11403 | 80 | |
XM_017011646.1 | XP_016867135.1 | 46 - 47 | 11418 | 80 | |
XM_017011647.1 | XP_016867136.1 | 46 - 47 | 11364 | 80 | |
XM_017011648.1 | XP_016867137.1 | 46 - 47 | 11361 | 80 | |
XM_017011649.1 | XP_016867138.1 | 46 - 47 | 11394 | 80 | |
XM_017011650.1 | XP_016867139.1 | 45 - 46 | 11322 | 80 | |
XM_017011651.1 | XP_016867140.1 | 46 - 47 | 11316 | 80 | |
XM_017011652.1 | XP_016867141.1 | 46 - 47 | 11268 | 80 | |
XM_017011653.1 | XP_016867142.1 | 44 - 45 | 11229 | 80 | |
XM_017011654.1 | XP_016867143.1 | 44 - 45 | 11181 | 80 | |
XM_017011655.1 | XP_016867144.1 | 47 - 48 | 11116 | 80 | |
XM_017011656.1 | XP_016867145.1 | 46 - 47 | 11046 | 80 | |
XM_017011657.1 | XP_016867146.1 | 32 - 33 | 6966 | 80 | |
XM_017011658.1 | XP_016867147.1 | 25 - 26 | 6328 | 80 | |
XM_017011659.1 | XP_016867148.1 | 24 - 25 | 6289 | 80 | |
XM_017011660.1 | XP_016867149.1 | 24 - 25 | 6289 | 80 | |
GenBank mRNA | AB018346.1 | - | 18 - 19 | 4493 | 80 |
AB019691.1 | - | 45 - 46 | 11151 | 80 | |
AF083037.1 | - | 38 - 39 | 10153 | 80 | |
AF091711.1 | - | 19 - 20 | 4634 | 80 | |
AJ131693.1 | - | 45 - 46 | 11314 | 80 | |
AK025160.1 | - | 5 - 6 | 933 | 80 | |
AK026444.1 | - | 7 - 8 | 2013 | 80 | |
AK074869.1 | - | 15 - 16 | 3193 | 80 | |
BC040932.1 | - | 4 - 5 | 688 | 80 |