Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
Chromosome Location
Chr.X: 131273506 - 131289457 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170961.1 NP_001164432.1 14 - 15 2904 106
NM_001170962.1 NP_001164433.1 13 - 14 2862 106
NM_001555.4 NP_001546.2 14 - 15 2889 106
XM_011531330.1 XP_011529632.1 15 - 16 2802 106
XM_011531333.1 XP_011529635.1 14 - 15 2904 106
XM_011531334.1 XP_011529636.1 14 - 15 2904 106
GenBank mRNA AF034198.1 - 13 - 14 2711 106
AK226008.1 - 14 - 15 2761 106
AK295438.1 - 5 - 6 940 106
AK307871.1 - 14 - 15 2765 106
BC063884.1 - 14 - 15 2725 106
Y10523.1 - 13 - 14 2686 106

View Full Product Details