The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321.2 | NP_001312.1 | 1 - 2 | 102 | 60 |
GenBank mRNA | AY349166.1 | - | 1 - 2 | 59 | 60 |
BC000992.2 | - | 1 - 2 | 50 | 60 | |
BX461295.2 | - | 1 - 2 | 124 | 60 |