The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020154.2 | NP_064539.1 | 3 - 4 | 508 | 147 |
GenBank mRNA | AF242729.1 | - | 3 - 4 | 545 | 147 |
AJ245874.1 | - | 3 - 4 | 530 | 147 | |
AJ250344.1 | - | 3 - 4 | 508 | 147 | |
AK314883.1 | - | 3 - 4 | 577 | 147 | |
AY358445.1 | - | 3 - 4 | 526 | 147 | |
BC012456.1 | - | 3 - 4 | 472 | 147 | |
BC104934.1 | - | 3 - 4 | 586 | 147 | |
BC104936.1 | - | 3 - 4 | 586 | 147 |