The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003701.3 | NP_003692.1 | 2 - 3 | 533 | 72 |
NM_033012.3 | NP_143026.1 | 4 - 5 | 694 | 72 | |
XM_011535280.2 | XP_011533582.1 | 2 - 3 | 238 | 72 | |
XM_017020802.1 | XP_016876291.1 | 2 - 3 | 340 | 72 | |
XM_017020803.1 | XP_016876292.1 | 2 - 3 | 187 | 72 | |
GenBank mRNA | AB037599.1 | - | 2 - 3 | 259 | 72 |
AB061227.1 | - | 3 - 4 | 341 | 72 | |
AB064268.1 | - | 3 - 4 | 248 | 72 | |
AB064269.1 | - | 2 - 3 | 464 | 72 | |
AB064270.1 | - | 3 - 4 | 402 | 72 | |
AF013171.1 | - | 1 - 2 | 168 | 72 | |
AF019047.1 | - | 2 - 3 | 512 | 72 | |
AF053712.1 | - | 2 - 3 | 568 | 72 | |
BC040889.1 | - | 4 - 5 | 694 | 72 | |
BC074823.2 | - | 2 - 3 | 414 | 72 | |
BC074890.2 | - | 2 - 3 | 414 | 72 | |
BC117286.1 | - | 2 - 3 | 416 | 72 | |
BC117288.1 | - | 2 - 3 | 416 | 72 |