Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
Chromosome Location
Chr.12: 107317168 - 107659642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018072.1 NP_001018082.1 1 - 1 608 68
XM_017018807.1 XP_016874296.1 1 - 1 1860 68
GenBank mRNA BC027931.1 - 1 - 1 608 68
BC146824.1 - 1 - 1 607 68

View Full Product Details