The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181716.2 | NP_859067.2 | 3 - 4 | 645 | 75 |
GenBank mRNA | AF514992.1 | - | 3 - 4 | 581 | 75 |
AY382899.1 | - | 1 - 2 | 619 | 75 | |
BC052604.1 | - | 3 - 4 | 655 | 75 | |
BC137486.1 | - | 3 - 4 | 677 | 75 | |
BC137487.1 | - | 3 - 4 | 677 | 75 | |
BU618545.1 | - | 2 - 3 | 75 |