Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP6
Entrez Gene ID
80320
Gene Name
Sp6 transcription factor
Gene Aliases
EPFN, EPIPROFIN, KLF14
Chromosome Location
Chr.17: 47844908 - 47876312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258248.1 NP_001245177.1 1 - 2 234 136
GenBank mRNA AK127707.1 - 1 - 2 277 136

View Full Product Details