Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location
Chr.5: 151411344 - 151556318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308150.1 NP_001295079.1 6 - 7 726 85
NM_001308151.1 NP_001295080.1 6 - 7 696 85
NM_078483.3 NP_510968.2 6 - 7 726 85
XM_005268386.1 XP_005268443.1 6 - 7 696 85
XM_006714759.3 XP_006714822.1 6 - 7 775 85
XM_011537580.2 XP_011535882.1 6 - 7 1416 85
XM_011537581.1 XP_011535883.1 7 - 8 844 85
XM_011537583.2 XP_011535885.1 7 - 8 5281 85
XM_011537584.2 XP_011535886.1 6 - 7 935 85
XM_011537585.1 XP_011535887.1 7 - 8 791 85
XM_011537586.2 XP_011535888.1 6 - 7 1539 85
XM_011537587.2 XP_011535889.1 6 - 7 795 85
XM_011537588.2 XP_011535890.1 6 - 7 639 85
XM_011537589.2 XP_011535891.1 6 - 7 1547 85
XM_011537590.1 XP_011535892.1 6 - 7 554 85
XM_011537591.1 XP_011535893.1 6 - 7 529 85
XM_011537592.2 XP_011535894.1 6 - 7 533 85
XM_011537593.2 XP_011535895.1 7 - 8 789 85
XM_011537594.1 XP_011535896.1 8 - 9 903 85
XM_011537595.2 XP_011535897.2 6 - 7 1422 85
XM_011537596.2 XP_011535898.2 5 - 6 508 85
XM_017009216.1 XP_016864705.1 6 - 7 622 85
XM_017009217.1 XP_016864706.1 6 - 7 726 85
XM_017009218.1 XP_016864707.1 5 - 6 508 85
XM_017009219.1 XP_016864708.1 6 - 7 776 85
XM_017009220.1 XP_016864709.1 6 - 7 696 85
GenBank mRNA AF516142.1 - 5 - 6 518 85
AK057340.1 - 6 - 7 696 85
AK309323.1 - 6 - 7 698 85
AY162213.1 - 6 - 7 631 85
AY227111.1 - 7 - 8 646 85
AY227113.1 - 6 - 7 602 85
BC136437.1 - 6 - 7 597 85
BC136438.1 - 6 - 7 597 85
BX537963.1 - 6 - 7 723 85
GU296670.1 - 5 - 6 503 85

View Full Product Details