Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE2
Entrez Gene ID
84501
Gene Name
spire type actin nucleation factor 2
Gene Aliases
Spir-2
Chromosome Location
Chr.16: 89817477 - 89871319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032451.1 NP_115827.1 12 - 13 1829 102
XM_011523379.1 XP_011521681.1 12 - 13 2595 102
XM_011523380.2 XP_011521682.1 13 - 14 1851 102
XM_011523381.1 XP_011521683.1 13 - 14 2675 102
XM_017023779.1 XP_016879268.1 12 - 13 1841 102
XM_017023780.1 XP_016879269.1 12 - 13 2006 102
XM_017023781.1 XP_016879270.1 11 - 12 1478 102
GenBank mRNA AB058735.1 - 10 - 11 1927 102
AJ422077.1 - 12 - 13 1819 102
AK094918.1 - 5 - 6 646 102
AK130685.1 - 2 - 3 1711 102
AL834408.1 - 12 - 13 1623 102
BC063706.1 - 4 - 5 442 102
BC139732.1 - 10 - 11 1447 102

View Full Product Details