The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130006.1 | NP_001123478.1 | 11 - 12 | 1671 | 86 |
NM_001130007.1 | NP_001123479.1 | 11 - 12 | 1695 | 86 | |
NM_002094.3 | NP_002085.2 | 11 - 12 | 1674 | 86 | |
GenBank mRNA | AK095483.1 | - | 11 - 12 | 1602 | 86 |
AK290382.1 | - | 9 - 10 | 1028 | 86 | |
AK315267.1 | - | 11 - 12 | 1115 | 86 | |
BC009503.2 | - | 11 - 12 | 1572 | 86 | |
BT006722.1 | - | 9 - 10 | 1009 | 86 | |
X17644.1 | - | 11 - 12 | 1660 | 86 |