The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282600.1 | NP_001269529.1 | 1 - 2 | 220 | 116 |
NM_001320810.1 | NP_001307739.1 | 1 - 2 | 220 | 116 | |
XM_017003406.1 | XP_016858895.1 | 1 - 2 | 310 | 116 | |
GenBank mRNA | AK295493.1 | - | 1 - 2 | 207 | 116 |
BI599815.1 | - | 1 - 2 | 224 | 116 | |
HQ589336.1 | - | 1 - 2 | 208 | 116 |