The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164883.1 | NP_001158355.1 | 13 - 14 | 2173 | 125 |
NM_001282597.2 | NP_001269526.1 | 13 - 14 | 2173 | 125 | |
NM_001282598.1 | NP_001269527.1 | 13 - 14 | 2026 | 125 | |
NM_001282599.1 | NP_001269528.1 | 7 - 8 | 1114 | 125 | |
NM_001282600.1 | NP_001269529.1 | 7 - 8 | 1057 | 125 | |
NM_001320810.1 | NP_001307739.1 | 7 - 8 | 1057 | 125 | |
NM_004389.3 | NP_004380.2 | 13 - 14 | 2173 | 125 | |
XM_011532555.2 | XP_011530857.1 | 13 - 14 | 2020 | 125 | |
XM_011532556.2 | XP_011530858.1 | 14 - 15 | 2216 | 125 | |
XM_011532557.2 | XP_011530859.1 | 5 - 6 | 762 | 125 | |
XM_017003403.1 | XP_016858892.1 | 14 - 15 | 2216 | 125 | |
XM_017003404.1 | XP_016858893.1 | 13 - 14 | 2111 | 125 | |
XM_017003405.1 | XP_016858894.1 | 13 - 14 | 2111 | 125 | |
XM_017003406.1 | XP_016858895.1 | 7 - 8 | 1147 | 125 | |
GenBank mRNA | AK127226.1 | - | 17 - 18 | 2619 | 125 |
AK295181.1 | - | 13 - 14 | 1972 | 125 | |
AK295493.1 | - | 7 - 8 | 1044 | 125 | |
AK303035.1 | - | 13 - 14 | 2026 | 125 | |
BC040458.1 | - | 12 - 13 | 1845 | 125 | |
BC052996.1 | - | 13 - 14 | 2107 | 125 | |
BX537769.1 | - | 7 - 8 | 1114 | 125 | |
HQ589336.1 | - | 7 - 8 | 1045 | 125 | |
M94151.2 | - | 12 - 13 | 1924 | 125 |