Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLD2
Entrez Gene ID
5338
Gene Name
phospholipase D2
Gene Aliases
-
Chromosome Location
Chr.17: 4807101 - 4823432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243108.1 NP_001230037.1 20 - 21 2247 74
NM_002663.4 NP_002654.3 20 - 21 2247 74
XM_005256696.2 XP_005256753.1 11 - 12 1307 74
XM_017024764.1 XP_016880253.1 20 - 21 2247 74
GenBank mRNA AB209374.1 - 15 - 16 3815 74
AF033850.1 - 20 - 21 2282 74
AF035483.1 - 19 - 20 2121 74
AF038440.1 - 20 - 21 2205 74
AF038441.1 - 20 - 21 2205 74
AK314797.1 - 20 - 21 2165 74
AY178289.1 - 11 - 12 1212 74
BC015033.1 - 20 - 21 2208 74
BC056871.1 - 20 - 21 2156 74

View Full Product Details