The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243108.1 | NP_001230037.1 | 23 - 24 | 2553 | 70 |
XM_017024764.1 | XP_016880253.1 | 23 - 24 | 2553 | 70 | |
GenBank mRNA | AF038441.1 | - | 23 - 24 | 2511 | 70 |