Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLD2
Entrez Gene ID
5338
Gene Name
phospholipase D2
Gene Aliases
-
Chromosome Location
Chr.17: 4807101 - 4823432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243108.1 NP_001230037.1 9 - 10 986 56
NM_002663.4 NP_002654.3 9 - 10 986 56
XM_017024764.1 XP_016880253.1 9 - 10 935 56
GenBank mRNA AB209374.1 - 6 - 7 1538 56
AF033850.1 - 9 - 10 1021 56
AF035483.1 - 8 - 9 860 56
AF038440.1 - 9 - 10 944 56
AF038441.1 - 9 - 10 944 56
AK304246.1 - 9 - 10 939 56
AK314797.1 - 9 - 10 904 56
BC015033.1 - 9 - 10 947 56
BC056871.1 - 9 - 10 895 56

View Full Product Details