The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142621.2 | NP_001136093.1 | 8 - 9 | 1764 | 70 |
NM_004257.5 | NP_004248.2 | 8 - 9 | 1767 | 70 | |
XM_011512193.2 | XP_011510495.1 | 8 - 9 | 1724 | 70 | |
XM_017005339.1 | XP_016860828.1 | 7 - 8 | 1260 | 70 | |
GenBank mRNA | AF022795.1 | - | 7 - 8 | 1702 | 70 |
AK291382.1 | - | 8 - 9 | 1732 | 70 | |
BC020548.2 | - | 8 - 9 | 1725 | 70 |