The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142286.1 | NP_001135758.1 | 24 - 25 | 3101 | 101 |
NM_024624.5 | NP_078900.1 | 23 - 24 | 3014 | 101 | |
XM_017004915.1 | XP_016860404.1 | 22 - 23 | 4334 | 101 | |
XM_017004916.1 | XP_016860405.1 | 22 - 23 | 2929 | 101 | |
GenBank mRNA | AJ310551.1 | - | 22 - 23 | 2805 | 101 |
AK025769.1 | - | 10 - 11 | 1410 | 101 | |
AK292421.1 | - | 27 - 28 | 3207 | 101 | |
AL136544.1 | - | 24 - 25 | 3075 | 101 | |
AL832979.1 | - | 23 - 24 | 2977 | 101 | |
BC039828.1 | - | 23 - 24 | 2998 | 101 |