The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142286.1 | NP_001135758.1 | 6 - 7 | 774 | 126 |
NM_024624.5 | NP_078900.1 | 5 - 6 | 687 | 126 | |
XM_011533107.2 | XP_011531409.1 | 4 - 5 | 2007 | 126 | |
XM_011533108.2 | XP_011531410.1 | 5 - 6 | 683 | 126 | |
XM_017004913.1 | XP_016860402.1 | 5 - 6 | 686 | 126 | |
XM_017004914.1 | XP_016860403.1 | 6 - 7 | 776 | 126 | |
XM_017004915.1 | XP_016860404.1 | 4 - 5 | 2007 | 126 | |
XM_017004916.1 | XP_016860405.1 | 5 - 6 | 683 | 126 | |
GenBank mRNA | AJ310551.1 | - | 4 - 5 | 478 | 126 |
AK092853.1 | - | 4 - 5 | 944 | 126 | |
AK098325.1 | - | 5 - 6 | 495 | 126 | |
AK292421.1 | - | 8 - 9 | 880 | 126 | |
AL136544.1 | - | 6 - 7 | 748 | 126 | |
AL832979.1 | - | 5 - 6 | 650 | 126 | |
BC032675.1 | - | 7 - 8 | 807 | 126 | |
BC039828.1 | - | 5 - 6 | 671 | 126 |