Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIPI2
Entrez Gene ID
26100
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
ATG18B, Atg21, CGI-50, WIPI-2
Chromosome Location
Chr.7: 5190204 - 5233855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033518.1 NP_001028690.1 8 - 9 954 98
NM_001033519.1 NP_001028691.1 7 - 8 900 98
NM_001033520.1 NP_001028692.1 5 - 6 793 98
NM_001278299.1 NP_001265228.1 8 - 9 1128 98
NM_015610.3 NP_056425.1 8 - 9 954 98
NM_016003.3 NP_057087.2 7 - 8 900 98
XM_006715685.1 XP_006715748.1 6 - 7 786 98
XM_017011952.1 XP_016867441.1 6 - 7 786 98
GenBank mRNA AF151808.1 - 5 - 6 792 98
AK023041.1 - 8 - 9 914 98
AK024279.1 - 7 - 8 879 98
AK315468.1 - 8 - 9 1074 98
AL080155.1 - 8 - 9 955 98
AY691425.1 - 7 - 8 684 98
AY691426.1 - 5 - 6 724 98
BC004116.2 - 8 - 9 897 98
BC007596.1 - 7 - 8 867 98
BC021012.1 - 7 - 8 819 98
BC021068.1 - 7 - 8 819 98
BC021200.2 - 8 - 9 895 98
CR533530.1 - 8 - 9 738 98

View Full Product Details