The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033518.1 | NP_001028690.1 | 9 - 10 | 1057 | 98 |
NM_001033519.1 | NP_001028691.1 | 8 - 9 | 1003 | 98 | |
NM_001033520.1 | NP_001028692.1 | 6 - 7 | 896 | 98 | |
NM_001278299.1 | NP_001265228.1 | 9 - 10 | 1231 | 98 | |
NM_015610.3 | NP_056425.1 | 9 - 10 | 1057 | 98 | |
NM_016003.3 | NP_057087.2 | 8 - 9 | 1003 | 98 | |
XM_006715685.1 | XP_006715748.1 | 7 - 8 | 889 | 98 | |
XM_017011952.1 | XP_016867441.1 | 7 - 8 | 889 | 98 | |
GenBank mRNA | AF151808.1 | - | 6 - 7 | 895 | 98 |
AK023041.1 | - | 9 - 10 | 1017 | 98 | |
AK024279.1 | - | 8 - 9 | 982 | 98 | |
AK315468.1 | - | 9 - 10 | 1177 | 98 | |
AL080155.1 | - | 9 - 10 | 1058 | 98 | |
AY691426.1 | - | 6 - 7 | 827 | 98 | |
BC004116.2 | - | 9 - 10 | 1000 | 98 | |
BC007596.1 | - | 8 - 9 | 970 | 98 | |
BC021012.1 | - | 8 - 9 | 922 | 98 | |
BC021068.1 | - | 8 - 9 | 922 | 98 | |
BC021200.2 | - | 9 - 10 | 998 | 98 | |
CR533530.1 | - | 9 - 10 | 841 | 98 |