The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015254.3 | NP_056069.2 | 25 - 26 | 3276 | 79 |
XM_005273458.1 | XP_005273515.1 | 25 - 26 | 3276 | 79 | |
XM_011544457.1 | XP_011542759.1 | 25 - 26 | 3276 | 79 | |
XM_011544458.1 | XP_011542760.1 | 25 - 26 | 3276 | 79 | |
XM_011544459.2 | XP_011542761.1 | 25 - 26 | 3261 | 79 | |
XM_011544460.2 | XP_011542762.1 | 25 - 26 | 3268 | 79 | |
XM_011544461.1 | XP_011542763.1 | 24 - 25 | 3263 | 79 | |
XM_017013257.1 | XP_016868746.1 | 25 - 26 | 3276 | 79 | |
GenBank mRNA | AB014539.3 | - | 25 - 26 | 3246 | 79 |
AF279865.1 | - | 25 - 26 | 3254 | 79 |