The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030649.2 | NP_085152.2 | 13 - 14 | 1090 | 61 |
XM_005244715.2 | XP_005244772.1 | 13 - 14 | 1126 | 61 | |
XM_011540606.2 | XP_011538908.1 | 13 - 14 | 1139 | 61 | |
XM_011540607.1 | XP_011538909.1 | 13 - 14 | 3563 | 61 | |
XM_011540608.2 | XP_011538910.1 | 12 - 13 | 1044 | 61 | |
XM_011540609.2 | XP_011538911.1 | 13 - 14 | 1140 | 61 | |
XM_017000233.1 | XP_016855722.1 | 13 - 14 | 1279 | 61 | |
GenBank mRNA | AB051503.1 | - | 12 - 13 | 2755 | 61 |
AF411981.1 | - | 11 - 12 | 890 | 61 | |
BC047001.1 | - | 3 - 4 | 542 | 61 | |
BC051194.1 | - | 13 - 14 | 1090 | 61 |