Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr.1: 1292384 - 1307925 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030649.2 NP_085152.2 23 - 24 2428 114
XM_005244715.2 XP_005244772.1 22 - 23 2365 114
XM_005244717.3 XP_005244774.1 11 - 12 1422 114
XM_011540606.2 XP_011538908.1 23 - 24 2477 114
XM_011540607.1 XP_011538909.1 23 - 24 4901 114
XM_011540608.2 XP_011538910.1 22 - 23 2382 114
XM_011540609.2 XP_011538911.1 22 - 23 2379 114
XM_017000233.1 XP_016855722.1 23 - 24 2617 114
GenBank mRNA AB051503.1 - 21 - 22 3994 114
AF411981.1 - 20 - 21 2129 114
BC004874.2 - 8 - 9 1032 114
BC021851.1 - 6 - 7 818 114
BC029929.1 - 3 - 4 231 114
BC047001.1 - 13 - 14 1880 114
BC051194.1 - 23 - 24 2428 114

View Full Product Details