Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr.1: 1292384 - 1307925 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030649.2 NP_085152.2 5 - 6 414 109
XM_005244715.2 XP_005244772.1 5 - 6 450 109
XM_011540606.2 XP_011538908.1 5 - 6 463 109
XM_011540607.1 XP_011538909.1 5 - 6 2887 109
XM_011540608.2 XP_011538910.1 5 - 6 464 109
XM_011540609.2 XP_011538911.1 5 - 6 464 109
GenBank mRNA AB051503.1 - 4 - 5 2079 109
AF411981.1 - 3 - 4 214 109
AK093954.1 - 5 - 6 461 109

View Full Product Details