The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030649.2 | NP_085152.2 | 7 - 8 | 639 | 59 |
XM_005244715.2 | XP_005244772.1 | 7 - 8 | 675 | 59 | |
XM_011540606.2 | XP_011538908.1 | 7 - 8 | 688 | 59 | |
XM_011540607.1 | XP_011538909.1 | 7 - 8 | 3112 | 59 | |
XM_011540609.2 | XP_011538911.1 | 7 - 8 | 689 | 59 | |
XM_017000233.1 | XP_016855722.1 | 7 - 8 | 828 | 59 | |
GenBank mRNA | AB051503.1 | - | 6 - 7 | 2304 | 59 |
AF411981.1 | - | 5 - 6 | 439 | 59 | |
AK093954.1 | - | 7 - 8 | 686 | 59 | |
BC051194.1 | - | 7 - 8 | 639 | 59 |