Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr.1: 1292384 - 1307925 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030649.2 NP_085152.2 1 - 2 120 89
XM_005244715.2 XP_005244772.1 1 - 2 156 89
XM_011540606.2 XP_011538908.1 1 - 2 139 89
XM_011540608.2 XP_011538910.1 1 - 2 140 89
XM_011540609.2 XP_011538911.1 1 - 2 140 89
XM_017000233.1 XP_016855722.1 1 - 2 71 89
GenBank mRNA BC051194.1 - 1 - 2 120 89

View Full Product Details