The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020971.2 | NP_066022.2 | 12 - 13 | 1764 | 68 |
XM_011527173.2 | XP_011525475.1 | 12 - 13 | 1830 | 68 | |
XM_017027049.1 | XP_016882538.1 | 12 - 13 | 2017 | 68 | |
XM_017027050.1 | XP_016882539.1 | 10 - 11 | 1399 | 68 | |
XM_017027051.1 | XP_016882540.1 | 12 - 13 | 1830 | 68 | |
XM_017027052.1 | XP_016882541.1 | 12 - 13 | 1830 | 68 | |
GenBank mRNA | AB046862.1 | - | 3 - 4 | 507 | 68 |
AF082075.1 | - | 12 - 13 | 1840 | 68 | |
AF311855.1 | - | 12 - 13 | 1764 | 68 | |
AF324063.1 | - | 12 - 13 | 1748 | 68 | |
AF324064.1 | - | 12 - 13 | 1748 | 68 | |
AY004226.1 | - | 11 - 12 | 1662 | 68 | |
AY004227.1 | - | 11 - 12 | 1662 | 68 |