Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 17 - 18 3955 60
XM_011527173.2 XP_011525475.1 17 - 18 4021 60
XM_017027049.1 XP_016882538.1 17 - 18 4208 60
XM_017027050.1 XP_016882539.1 15 - 16 3590 60
XM_017027051.1 XP_016882540.1 17 - 18 4021 60
GenBank mRNA AB046862.1 - 8 - 9 2698 60
AF082075.1 - 17 - 18 4016 60
AF311855.1 - 17 - 18 3955 60
AF324063.1 - 17 - 18 3939 60
AF324064.1 - 17 - 18 3939 60
AY004227.1 - 16 - 17 3838 60

View Full Product Details