Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 19 - 20 4197 62
NM_025213.2 NP_079489.2 2 - 3 250 62
XM_011527173.2 XP_011525475.1 19 - 20 4263 62
XM_017027049.1 XP_016882538.1 19 - 20 4450 62
XM_017027050.1 XP_016882539.1 17 - 18 3832 62
XM_017027051.1 XP_016882540.1 19 - 20 4263 62
GenBank mRNA AB046862.1 - 10 - 11 2940 62
AF082075.1 - 19 - 20 4258 62
AF311855.1 - 19 - 20 4197 62
AF311856.1 - 2 - 3 250 62
AF324063.1 - 19 - 20 4181 62
AF324064.1 - 19 - 20 4181 62
AK308547.1 - 2 - 3 238 62
AK309418.1 - 2 - 3 212 62
AY004226.1 - 19 - 20 4166 62
AY004227.1 - 18 - 19 4080 62
DA514440.1 - 2 - 3 212 62

View Full Product Details