Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 29 - 30 6243 65
XM_011527173.2 XP_011525475.1 29 - 30 6309 65
XM_017027049.1 XP_016882538.1 29 - 30 6496 65
XM_017027050.1 XP_016882539.1 27 - 28 5878 65
XM_017027051.1 XP_016882540.1 29 - 30 6309 65
GenBank mRNA AB046862.1 - 20 - 21 4986 65
AF082075.1 - 29 - 30 6304 65
AF311855.1 - 29 - 30 6243 65
AF324063.1 - 29 - 30 6227 65
AY004226.1 - 29 - 30 6212 65
AY004227.1 - 28 - 29 6126 65

View Full Product Details