Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 31 - 32 7060 74
XM_011527173.2 XP_011525475.1 31 - 32 7126 74
XM_017027049.1 XP_016882538.1 31 - 32 7313 74
XM_017027050.1 XP_016882539.1 29 - 30 6695 74
GenBank mRNA AB046862.1 - 22 - 23 5803 74
AF082075.1 - 31 - 32 7121 74
AF311855.1 - 31 - 32 7060 74
AF324063.1 - 31 - 32 7044 74
AY004226.1 - 31 - 32 7029 74

View Full Product Details