The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006784.2 | NP_006775.1 | 9 - 10 | 1061 | 172 |
GenBank mRNA | AF083217.1 | - | 9 - 10 | 1042 | 172 |
AK292438.1 | - | 9 - 10 | 1056 | 172 |